Polyethylene glycol diacrylate (PEGDA), M.W. 8,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate
CAS Number:26570-48-9
:
Formula:CH₂=COC(OCH₂CH₂)nOCOC=CH₂
MDL Number:MFCD00081876
EINECS:607-960-5
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together