Order Entry
United Kingdom
ContactUsLinkComponent
poly(3-hexylthiophene-2,5-diyl) regioregular low metals
poly(3-hexylthiophene-2,5-diyl) regioregular low metals
Catalog # 44745.02
CAS Number:  
poly(3-hexylthiophene-2,5-diyl) regioregular low metals
Catalog # 44745.02
CAS Number:  

Specifications

  • Pk:
    0,5 g
  • Environmentally Preferable:

Specifications

About this item

Synonyms: P3HT, Poly(3-hexylthiophene), Poly[3-hexylthiophene-2,5-diyl]
CAS Number:104934-50-1
 :
Formula:C₁₂H₂₀S
MDL Number:MFCD00217686
 :GHS07
Storage Temperature:Ambient
 :110134-47-9