Cell counter, Cellometer® K2
The Cellometer K2 is a cell imager for automated dual-channel fluorescent image capture and analysis.
- Analyse clumpy and irregular-shaped cells
- Count and viability measurements ten times faster than manual methods
- Disposable counting chamber, no cross-contamination
- Automatically generates a cell size histogram based on cell diameter
- Available fluorescence optics: Modules excitation/emission (470/535 nm) and excitation/emission (540/660 nm)
Cells lines and primary cells in complex samples are characterized by heavy debris, red blood cell contamination, low cell concentrations and multiple cell types including PBMCs, stem cells, cancer cells, splenocytes, hepatocytes and blood.
The K2 can perform cell count, size, viability, intensity and population analysis as well as cell-based assays such as apoptosis, cell cycle and GFP analysis.
: Includes a Cellometer software, two fluorescence modules, a power supply, USB 2.0 connection cable and phone/online applications support during set-up.
- Catalog No:
- NEXCMK2
- NEXCCMK2-SK150-FCS
- Concentration:
- 105 - 107 cells/ml
- 105 - 107 cells/ml
- Cell size:
- 5 − 300 µm
- 5 − 300 µm
- Electrical:
- 100 – 240 VAC; 50/60 Hz; 1,0 A (input to adapter) – 12 VDC; 3,34 A (output to instrument)
- 100 – 240 VAC; 50/60 Hz; 1,0 A (input to adapter) – 12 VDC; 3,34 A (output to instrument)
- W×D×H:
- 152×216×356 mm
- 152×216×356 mm
- Weight:
- 10,4 kg
- 10,4 kg
Frequently Bought Together