Dishes, crystallising
Supplier: MARIENFELD
Certificates
About this item
Crystallising dishes, made of borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance up to +500 °C
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Cylindrical shape with fused rim and flat bottom.
Frequently Bought Together