Order Entry
United Kingdom
ContactUsLinkComponent
Magnetic Beads, Sera-Mag SpeedBeads Streptavidin-blocked magnetic particles
Magnetic Beads, Sera-Mag SpeedBeads Streptavidin-blocked magnetic particles
Supplier:  Cytiva
Certificates
undefined
Magnetic Beads, Sera-Mag SpeedBeads Streptavidin-blocked magnetic particles
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • 21152104010350.
  • 21152104011150.
  • 21152104010150.
  • Binding capacity:
  • 3000 - 4000 pmole/mg beads
  • 3000 - 4000 pmole/mg beads
  • 3000 - 4000 pmole/mg beads
  • Particle size:
  • nominal 1 µm diameter
  • nominal 1 µm diameter
  • nominal 1 µm diameter