Order Entry
United Kingdom
ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) solution for the detection of chlorine, Supelco®
3,3',5,5'-Tetramethylbenzidine (TMB) solution for the detection of chlorine, Supelco®
Catalog # 1.08475.1000
Supplier:  Merck
CAS Number:  
undefined
3,3',5,5'-Tetramethylbenzidine (TMB) solution for the detection of chlorine, Supelco®
Catalog # 1.08475.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.023 g/cm3
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material
  • For:
    Detection of chlorine

Specification Test Results

About this item

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
 :
 :240.35 g/mol
UN:3264
 :GHS05
Storage Temperature:Ambient
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748
Boiling Pt:402
ADR:8 II
 :M-Clarity™ quality level = MQ100