Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) 75% ≥75% in 2-propanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Diisopropoxytitanium(IV) Bis(acetylacetonate), Titanium (di-i-propoxide)bis(acetylacetonate), Titanium(IV) Diisopropoxide Bis(acetylacetonate), Titanium (IV) bis (acetylacetonate) diisopropoxide
CAS Number:17927-72-9
:
Formula:C₁₆H₂₈O₆Ti
MDL Number:MFCD00000031
Molecular Weight:364.26 g/mol
ADR:3 II
:M-Clarity™ quality level = MQ200
UN:1219
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) Hydrolysis
- Flash Point:12 °C
- Density:1.01 g/cm3 (20 °C)
- Assay (ex Ti):72,0 - 78,0%
- Density (d 20 °C/ 4 °C):0,990 - 1,020
- Identity (IR):passes test
Specification Test Results
Frequently Bought Together