Anti-STM Rabbit Polyclonal Antibody
AGRIAS111764
: Agrisera
- Antibody type:Primary
- Antigen symbol:STM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:A. thaliana
- Western blot:Yes
- Environmentally Preferable:
- Immunogen:KLH-conjugated peptide chosen from Arabidopsis thaliana STM protein sequence, UniProt: Q38874,TAIR: At1g62360
- Size:50 µg
- Pk:50 µG
Homeobox protein SHOOT MERISTEMLESS is a protein required for shoot apical meristem (SAM) formation during embryogenesis.,Homeobox protein SHOOT MERISTEMLESS is a protein required for shoot apical meristem (SAM) formation during embryogenesis.
Type: Primary
Antigen: STM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana