Anti-ALKBH8 Rabbit Polyclonal Antibody
BOSSBS-6510R
: Bioss
- Antibody type:Primary
- Antigen name:AlkB Homolog 8, TRNA Methyltransferase
- Antigen symbol:ALKBH8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:91801
- Antigen synonyms:ABH8|TRM9|TRMT9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Catalyzes the methylation of 5-carboxymethyl uridine to 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in tRNA. Catalyzes the last step in the formation of 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in target tRNA. Has a preference for tRNA(Arg) and tRNA(Glu), and does not bind tRNA(Lys). Required for normal survival after DNA damage. May inhibit apoptosis and promote cell survival and angiogenesis.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ALKBH8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat