Anti-ADCY9 Rabbit Polyclonal Antibody
BOSSBS-3926R
: Bioss
- Antibody type:Primary
- Antigen name:Adenylate Cyclase 9
- Antigen symbol:ADCY9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:115
- Antigen synonyms:Adenylate cyclase type IX|Adenylyl cyclase 9|ADCY9|AC9|ATP pyrophosphate-lyase 9|KIAA0520|Adenylate cyclase type 9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:260-280/1353
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
May play a fundamental role in situations where fine interplay between intracellular calcium and cAMP determines the cellular function. May be a physiologically relevant docking site for calcineurin (By similarity).
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ADCY9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat