Anti-ADCY5 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3922R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Adenylate Cyclase 5
- Antigen symbol:ADCY5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:111
- Antigen synonyms:ADCY5|AC5|Adenylate cyclase type V|FDFM|Adenylate cyclase type 5|ATP pyrophosphate-lyase 5|Adenylyl cyclase 5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:860-909/1261
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
This is a membrane-bound, calcium-inhibitable adenylyl cyclase.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ADCY5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat