- Antibody type:Primary
- Antigen name:profilin 1
- Antigen symbol:PFN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:5216
- Antigen synonyms:ALS18
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Profilin 1 is a ubiquitous actin monomer-binding protein involved in actin polymerization in response to extracellular signals. Three profilin genes have been identified: profilin 1, 2, and 3. Profilin 1 is the most ubiquitous and abundant, and is highly expressed throughout development and adulthood inmost tissues including brain. Profilin 2 is the neuronal specific isoform and profilin 3 is a testis specific isoform. Profilins of eukaryotic cells are small (12-15 kDa) cytoplasmic proteins that bind to actin monomers, polyphosphoinositides and polymers of L-proline. Profilins were shown to be important for normal cell proliferation, differentiation and motility. Deletion of profilin 1 gene leads to an embryonic lethal phenotype.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Profilin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat