Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HELZ Rabbit Polyclonal Antibody
Anti-HELZ Rabbit Polyclonal Antibody
Catalog # BOSSBS-15450R
Supplier:  Bioss
Anti-HELZ Rabbit Polyclonal Antibody
Catalog # BOSSBS-15450R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HELZ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Antigen synonyms:
    Helicase with zinc finger|HUMORF5|KIAA0054|HUMORF 5|HELZ_HUMAN.|Down regulated in human cancers|MGC163454|DRHC|Probable helicase with zinc finger domain|DKFZp586G1924
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Helicases comprise a class of enzymes that function as motor proteins which move along nucleic acid phosphodiester bonds, effectively separating two annealed nucleic acid strands. RNA helicases alter the conformation of RNA, specifically by unwinding double-stranded RNA regions to yield single RNA strands, a process which changes the biological activity of the RNA molecule. HELZ (helicase with zinc finger), also known as DHRC or HUMORF5, is a 1,942 amino acid nuclear protein that contains one C3H1-type zinc finger and belongs to the RNA helicase superfamily. Expressed ubiquitously during embryonic development, HELZ is thought to function as an RNA helicase that modifies RNA structure and plays a role in the development of multiple organs and tissues within the developing embryo.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HELZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat