Anti-Neurotensin Receptor 1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-12002R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Neurotensin Receptor 1
- Antigen symbol:NTR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:4923
- Antigen synonyms:NTRH|NTRR|High-affinity levocabastine-insensitive neurotensin receptor|NT-R-1|NTSR1|NTR1|Neurotensin receptor type 1|NTR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:210-234/418
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
G-protein coupled receptor for the tridecapeptide neurotensin (NTS). Signaling is effected via G proteins that activate a phosphatidylinositol-calcium second messenger system. Signaling leads to the activation of downstream MAP kinases and protects cells against apoptosis.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: Neurotensin Receptor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human