Specifications
- Antibody type:Primary
- Antigen name:Bromodomain and PHD Finger-containing Transcription Factor
- Antigen symbol:BPTF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:bromodomain and PHD finger-containing transcription factor|NURF301|FAC1|large subunit|nucleosome remodeling factor|fetal Alz-50 clone 1 protein|fetal Alzheimer antigen|bromodomain and PHD domain transcription factor|fetal Alz-50 reactive clone 1|FALZ|nucleosome-remodeling factor subunit BPTF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
BPTF is a 2,907 amino acid protein encoded by the human gene BPTF. BPTF belongs to the PBTF family and contains one bromodomain, one DDT domain and two PHD-type zinc fingers. BPTF acts as a histone-binding component of NURF (nucleosome-remodeling factor). The NURF complex, which consists of SMARCA1, BPTF, RbAp46 and RbAp48, acts to catalyze ATP-dependent nucleosome sliding and facilitates transcription of chromatin. It specifically recognizes histone H3 tails trimethylated on 'Lys-4' (H3-K4Me3), which mark transcription start sites of virtually all active genes. BPTF may also help regulate transcription through direct binding to DNA or transcription factors.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: BPTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat