Anti-CIDEB Rabbit Polyclonal Antibody
BOSSBS-0544R
: Bioss
- Antibody type:Primary
- Antigen name:Cell death-inducing DFFA-like effector B
- Antigen symbol:CIDEB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Gene ID:27141
- Antigen synonyms:CIDEB_HUMAN.|Cell death activator CIDE B|Cell death inducing DFFA like effector B|Cell death activator CIDE-B|CIDEB|Cell death-inducing DFFA-like effector B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:100-150/219
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Activates apoptosis.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CIDEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: