Anti-LIMA1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB29449
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:LIM domain and actin binding 1
- Antigen symbol:LIMA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:51474
- Antigen synonyms:SREBP3|EPLIN
- Sequence:RPHKDLWASKNENEEILERPAQLANARETPHSPGVEDAPIAKVGVLAASMEAKASSQQEKEDKPAETKKLRIAWPPPTELGSSGSALEEGI
- Immunogen:Recombinant protein corresponding to human LIMA1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant human LIMA1.
Type: Primary
Antigen: LIMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human