Bottle/tube rollers
444-0960
- Shaking motion:Rolling
- Description:Bottle/tube roller, 100 - 240 V, 50/60 Hz
- Environmentally Preferable:
- Max. load:6,5 kg
- Control:PID control
- Input power:10 W
- No. of rollers:6
- Operating temperature:4...60 °C
- Roller size Lר:259×28 mm
- Speed:1 - 80 min⁻¹
- Timer:Continuous or 1 min to 99 h 59 min
- Weight:6,5 kg
- W×D×H:378×360×72 mm
- Pk:1
This sleek compact design unit is capable of rolling multiple size tubes or bottles for applications like cell culture, hybridisation assays and liquid mixing. The anti-corrosive, wear-resistant construction enables use in humid and CO₂ environments.
- Adjustable speed accommodates a variety of applications
- Adjustable rollers accommodate a variety of tube/bottle sizes
- Stacking kit allows two units to be stacked, saving valuable bench space
- Optional remote control facilitates operating unit while in incubator and adds timer function
: cCSAus compliance.
: Includes O-rings to ensure smooth rolling when using tubes. Bars must be ordered seperately.