Anti-Flt3 ligand Antibody
Catalog # BSBTPB9588
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Fms-related tyrosine kinase 3 ligand
- Antigen symbol:FLT3LG
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2323
- Antigen synonyms:FLT3L|FL
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Fms-related tyrosine kinase 3 ligand(FLT3LG) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: Flt3 ligand
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat