Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Apolipoprotein J Antibody
Anti-Apolipoprotein J Antibody
Catalog # BSBTPB9575
Supplier:  Boster Bio
undefined
Anti-Apolipoprotein J Antibody
Catalog # BSBTPB9575
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    clusterin
  • Antigen symbol:
    CLU
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    1191
  • Antigen synonyms:
    SGP-2|NA1/NA2|KUB1|SP-40|TRPM-2|SGP2|CLU1|APOJ|CLI|APO-J|TRPM2|AAG4|CLU2
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Clusterin(CLU) detection. Tested with WB, IHC-P, ELISA in Human.

Type: Primary
Antigen: Apolipoprotein J
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human