Order Entry
United Kingdom
ContactUsLinkComponent
NBB®-C, Concentrated Broth
NBB®-C, Concentrated Broth
Catalog # 710-0851
Supplier:  DOEHLER GROUP
CAS Number:  
undefined
NBB®-C, Concentrated Broth
Catalog # 710-0851
Supplier:  DOEHLER GROUP
CAS Number:  
Specifications
  • Pack type:
    Bottle
  • Pk:
    9
  • Standards:
    -
  • Description:
    NBB®-concentrate (NBB®-C), 250 ml
  • Environmentally Preferable:
  • Filling:
    250 ml
Specifications
About this item
Concentrated, selective liquid medium for fast trace detection in unfiltered beer. Under its NBB® brand, Döhler offers innovative culture media for the fast and reliable detection of beverage spoiling microorganisms, such as Lactobacilli, Pectinatus and Megasphaera, in all samples that arise in breweries, as well as for the wine industry. NBB®-C is a liquid culture medium with enhanced selectivity through the addition of yeast-cloudy beer samples.
  • Simply added to the beer sample for analysis Final microscopic inspection, no indicator verification