Intestinal villi model
BINH1005532
- Description:intestinal villi model
- Environmentally Preferable:
- Pk:1
This intestinal villi model consists of one entire villus, one longitudinally sectioned villus showing the arterioles and venules and one sectioned villus to show the lymphatic vessels. The intestinal villi model also includes a longitudinal section of Lieberkühn's crypt.
- Magnified 100 times
- On base