Peqlab peqGOLD, Protein Marker V (Pre-Stained)
Catalog # 27-2210
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 KIT
- Description:Protein Marker V (pre-stained, ready to use), 2×250 µl, sufficient for 50 lanes
- Environmentally Preferable:
Specifications
About this item
Protein Marker V (pre-stained) for monitoring protein separation, transfer efficiency and size determination.
- Mix of nine recombinant proteins
- Covalently pre-stained by using one of three different dyes
- Range: 10 to 250 kDa
- Concentration: 100 to 200 ng/µl each protein
- Ready to use for a fast, convenient workflow