Anti-G-CSF Rabbit Polyclonal Antibody
BSBTPB9563
- Antibody type:Primary
- Antigen name:colony stimulating factor 3 (granulocyte)
- Antigen symbol:CSF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1440
- Antigen synonyms:C17orf33|GCSF|CSF3OS
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Granulocyte colony-stimulating factor(CSF3) detection. Tested with WB, ELISA in Human.
Type: Primary
Antigen: G-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human