Anti-Parvin alpha Rabbit Polyclonal Antibody
Catalog # BSBTPB9555
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:parvin, alpha
- Antigen symbol:PARVA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55742
- Antigen synonyms:MXRA2|CH-ILKBP
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Alpha-parvin(PARVA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: Parvin alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat