Peqlab peqGOLD, DNA Ladder, 100 bp
Catalog # 25-2010
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 KIT
- Description:DNA ladder, 100 bp (concentrated), 500 ng/µl, 1×100 µl, sufficient for 50 lanes
- Environmentally Preferable:
Specifications
About this item
DNA ladder for quantitative and qualitative analysis of dsDNA.
- Range: 10 fragments, 100 to 1000 bp in size
- Highlighted 500 bp band for easy orientation
- Also available as ready to use mix containing Orange G dye instead of bromophenol blue