Anti-PPP2R2B Antibody Pair
ABNOH00005521-PW1
: Abnova
- Antigen name:Protein Phosphatase 2, Regulatory Subunit 2B
- Antigen symbol:PPP2R2B
- Host:Multiple
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:5521
- Antigen synonyms:Serine/threonine protein phosphatase 2A 55 kDa regulatory subunit B beta isoform|PP2A subunit B isoform beta|PP2A PR55B|Protein phosphatase 2 regulatory subunit B|Serine/threonine protein phosphatase 2A neuronal isoform|PP2A subunit B isoform B55 beta|Protein phosphatase 2 regulatory subunit B beta isoform|PP2A subunit B PR55 beta isoform|PR2AB55 beta|PR2APR55 beta|SCA12|2ABB_HUMAN|Serine/Threonine Protein Phosphatase 2A 56kDa Regulatory Subunit Alpha Isoform|PP2A subunit B isoform B55-beta|Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform.|Protein phosphatase 2 formerly 2A regulatory subunit B PR 52 beta isoform|PP2A B55beta|SCA 12|PP2A subunit B B beta isoform|Ppp2r2b|PP2A subunit B B55 beta isoform|Beta isoform of regulatory subunit B55 protein phosphatase 2|PP2A subunit B isoform R2 beta|PR52B|PP2A subunit B isoform PR55-beta|PP2A subunit B R2 beta isoform|PP2A subunit B isoform R2-beta|B55 beta|PR2AB beta|PP2A B Subunit B Alpha Isoform|PP2A subunit B isoform PR55 beta|MGC24888|PR55 beta|Protein phosphatase 2 formerly 2A regulatory subunit B beta isoform
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-PPP2R2B (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-PPP2R2B (50 µl)
Type:
Antigen: PPP2R2B
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human