Anti-CDK2 Antibody Pair
Catalog # ABNOH00001017-PW1
Supplier: Abnova
Specifications
- Antigen name:Cyclin-dependent kinase 2
- Antigen symbol:CDK2
- Host:Multiple
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:1017
- Antigen synonyms:Cell division protein kinase 2|CDH2 T160,Cyclin-dependent kinase 2|p-CDH2 Thr160|CDK2_HUMAN.|p33 protein kinase|CDH2 Thr160
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-CDK2 (300 µl); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-CDK2 (50 µl)
Type:
Antigen: CDK2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human