Anti-AMT Rabbit Polyclonal Antibody
ABNOH00000275-PW1
: Abnova
- Antigen name:Aminomethyltransferase
- Antigen symbol:AMT
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:275
- Antigen synonyms:GCST|NKH|GCVT|GCE|AMT|Glycine cleavage system T protein
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-AMT (300 ?l); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-AMT (50 ?l)
Type:
Antigen: AMT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human