Order Entry
United Kingdom
ContactUsLinkComponent
Anti-DGKZ Polyclonal Antibody Pair
Anti-DGKZ Polyclonal Antibody Pair
Catalog # ABNOH00008525-AP21
Supplier:  Abnova
Anti-DGKZ Polyclonal Antibody Pair
Catalog # ABNOH00008525-AP21
Supplier:  Abnova

Specifications

  • Antigen name:
    Diacylglycerol kinase, zeta
  • Antigen symbol:
    DGKZ
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    8525
  • Antigen synonyms:
    DAGK6|DAG kinase zeta|DGK-zeta|Diacylglycerol kinase|zeta|Diglyceride kinase
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Set comes with matched Antibody Pair to detect and quantify protein level of human DGKZ.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-DGKZ (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-DGKZ (20 µg);

Type:
Antigen: DGKZ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human