Anti-HMGB1 Antibody Pair
ABNOH00003146-PW1
: Abnova
- Antigen name:High-mobility Group Box 1
- Antigen symbol:HMGB1
- Host:Multiple
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:3146
- Antigen synonyms:high-mobility group box 1|RP11-550P23.1|high mobility group protein 1|Amphoterin|SBP-1|high-mobility group (nonhistone chromosomal) protein 1|high mobility group protein B1|HMG3|Sulfoglucuronyl carbohydrate binding protein|HMG1|HMG-1
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-HMGB1 (300 µl); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-HMGB1 (50 µg)
Type:
Antigen: HMGB1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human