Anti-CPNE1 Polyclonal Antibody Pair
ABNOH00008904-AP21
: Abnova
- Antigen name:Copine 1
- Antigen symbol:CPNE1
- Clonality:Polyclonal
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:8904
- Antigen synonyms:CPN1|COPN1
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Set comes with matched Antibody Pair to detect and quantify protein level of human CPNE1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-CPNE1 (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-CPNE1 (40 µl);
Type:
Antigen: CPNE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human