Anti-YAP1 Antibody Pair
Catalog # ABNOH00010413-AP11
Supplier: Abnova
Specifications
- Antigen name:Yes-associated protein 1
- Antigen symbol:YAP1
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:10413
- Antigen synonyms:65kDa|YAP|yorkie homolog|yes-associated protein 2|YAP2|Yes-associated protein 1|65 kDa Yes-associated protein|YKI|YAP65|yes-associated protein delta|yes-associated protein beta
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human YAP1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-YAP1 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-YAP1, IgG2a kappa (20 µg);
Type:
Antigen: YAP1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human