Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-STBD1 Antibody Pair
  ABNOH00008987-AP51
 :  Abnova
Anti-STBD1 Antibody Pair
  ABNOH00008987-AP51
 :  Abnova

 

  • Antigen name:
    Starch binding domain 1
  • Antigen symbol:
    STBD1
  • Host:
    Multiple
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    8987
  • Antigen synonyms:
    GENX-3414|Glycophagy Cargo Receptor STBD1|Genethonin 1|GENEX3414
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Set comes with matched Antibody Pair to detect and quantify protein level of human STBD1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-STBD1 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-STBD1 (50 µg);

Type:
Antigen: STBD1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human