Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-GRAP2 + ERBB2 Antibody Pair
  ABNODI0167
 :  Abnova
Anti-GRAP2 + ERBB2 Antibody Pair
  ABNODI0167
 :  Abnova

 

  • Antigen symbol:
    GRAP2 + ERBB2
  • Host:
    Multiple
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Gene ID:
    9402|2064
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the GRAP2 protein, and the other against the ERBB2 protein for use in Proximity Ligation Assay

Antibody pair set content: 1. GRAP2 Rabbit purified Polyclonal Antibody (20 µg) 2. ERBB2 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: GRAP2 + ERBB2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: