Anti-BAD Antibody Pair
ABNODP0092
: Abnova
- Antigen name:BCL2-associated agonist of cell death
- Antigen symbol:BAD
- Host:Multiple
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Gene ID:572
- Antigen synonyms:BCL2L8|Bcl-2-binding component 6|BBC6|Bcl2 antagonist of cell death|BBC2|BAD
- Modification:phospho-Ser134
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the BAD protein, and the other against the specific S134 phosphorylated site of BAD for use in Proximity Ligation Assay
Antibody pair set content: 1. Phospho-BAD S134 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. BAD Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: BAD
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: