Order Entry
United Kingdom
ContactUsLinkComponent
Rotavac Vacuum Pumps
undefined
Rotavac Vacuum Pumps
  • Catalog No:
  • HEID591-00141-03
  • HEID591-00160-03
  • 531-0457
  • HEID591-00142-00
  • Model:
  • Rotavac Vario Control
  • Rotavac Valve Tec
  • Rotavac Valve Tec
  • Rotavac Vario
  • Flow rate:
  • 1,7 m³/h
  • 0,75 m³/h
  • 0,75 m³/h
  • 1,7 m³/h
  • Weight:
  • 5,4 kg
  • 6 kg
  • 6 kg
  • 6 kg
  • W×D×H:
  • 236×167×196 mm
  • 316×183×143 mm (230 V version); 323×183×143 mm (120 V version)
  • 316×183×143 mm (230 V version); 323×183×143 mm (120 V version)
  • 193×263×299 mm
  • Power input:
  • 160
  • 80
  • 80
  • 160
  • Ultimate vacuum:
  • 2 mbar
  • 12 mbar
  • 12 mbar
  • 5 mbar
Frequently Bought Together