DNA ladder, 500 bp, DNAmark™
Catalog # GENO786-462
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 KIT
- Description:DNAmark™ 500 bp ladder, 50 µg
- Seq:04
- Environmentally Preferable:
Specifications
About this item
DNAmark™ 500 bp ladder consists of 10 DNA fragments ranging from 500 bp to 5000 bp. The 2000 bp fragment is present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 2000 bp are 40 ng. The 2000 bp fragment is 100 ng.
- Pre-mixed with loading dye and ready to use