Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-SGCA Rabbit Polyclonal Antibody
  BSBTPB9503
undefined
Anti-SGCA Rabbit Polyclonal Antibody
  BSBTPB9503

 

  • Antibody type:
    Primary
  • Antigen name:
    Sarcoglycan alpha (50kDa dystrophin-associated glycoprotein)
  • Antigen symbol:
    SGCA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    6442
  • Antigen synonyms:
    ADL|DAG2|DMDA2|50-DAG|adhalin|SCARMD1|A2|LGMD2D
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Alpha-sarcoglycan(SGCA) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: SGCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat