DNA ladder
Supplier: LONZA
Certificates
About this item
Thse DNA ladders provides rapid and easy size determination of the DNA samples. Each band has an ideal 50 % G-C content for precise sizing and sharp resolution. It is free from contaminating plasmid DNA with minimal background.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together