Order Entry
United Kingdom
ContactUsLinkComponent
Anti-AQP3 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Anti-AQP3 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ABNOPAB28886
Supplier:  Abnova
Anti-AQP3 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ABNOPAB28886
Supplier:  Abnova

Specifications

  • Antigen name:
    Aquaporin 3
  • Antigen symbol:
    AQP3
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    65133
  • Antigen synonyms:
    Aquaporin-3|Aquaglyceroporin-3|Aquaporin 3|GIL|AQP 3|AQP-3
  • Storage buffer:
    PBS pH 7,4, 50% glycerol and 0,09% sodium azide
  • Sequence:
    CHLEQPPPSTEAENVKLAHMKHKEQI
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to Rat Aqp3 C-terminus.
  • Purification:
    Affinity purification
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against recombinant Rat Aqp3.

Recommended Dilutions: Immunofluorescence: 1:200-400; Western Blot: 1:1000-4000

Type:
Antigen: AQP3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: