Anti-EGF Mouse Monoclonal Antibody [clone: 4E11]
Catalog # ABNOMAB10631
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Epidermal growth factor
- Antigen symbol:EGF
- Clonality:Monoclonal
- Clone:4E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:1950
- Antigen synonyms:kinesin-related motor protein Eg5|thyroid receptor-interacting protein 5|TRIP5|TR-interacting protein 5|kinesin-like spindle protein HKSP|HKSP|kinesin-like 1|thyroid receptor interacting protein 5|kinesin-like protein 1|KNSL1|TRIP-5|kinesin-like protein KIF11
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human EGF.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant EGF.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000
Type: Primary
Antigen: EGF
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human