Order Entry
United Kingdom
ContactUsLinkComponent
Anti-FUT4 Mouse Monoclonal Antibody [clone: 4E10]
Anti-FUT4 Mouse Monoclonal Antibody [clone: 4E10]
Catalog # ABNOMAB10526
Supplier:  Abnova
Anti-FUT4 Mouse Monoclonal Antibody [clone: 4E10]
Catalog # ABNOMAB10526
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fucosyltransferase 4
  • Antigen symbol:
    FUT4
  • Clonality:
    Monoclonal
  • Clone:
    4E10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    2526
  • Antigen synonyms:
    CD15|FUC-TIV|ELFT|SSEA-1|LeX|FCT3A|FUTIV
  • Storage buffer:
    0,03% sodium azide
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human FUT4.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of FUT4.

Recommended Dilutions: ELISA: 1:10000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: 4E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human