Order Entry
United Kingdom
ContactUsLinkComponent
Anti-APITD1 Mouse Monoclonal Antibody [clone: 6F16]
Anti-APITD1 Mouse Monoclonal Antibody [clone: 6F16]
Catalog # ABNOH00378708-M07
Supplier:  Abnova
Anti-APITD1 Mouse Monoclonal Antibody [clone: 6F16]
Catalog # ABNOH00378708-M07
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Apoptosis-Inducing, TAF9-Like Domain 1
  • Antigen symbol:
    APITD1
  • Clonality:
    Monoclonal
  • Clone:
    6F16
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    378708
  • Antigen synonyms:
    CENPS|CENP-S|FAAP16|MHF1
  • Amino acid number:
    1 to 138
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    MEEEAETEEQQRFSYQQRLKAAVHYTVGCLCEEVALDKEMQFSKQTIAAISELTFRQCENFAKDLEMFARHAKRTTINTEDVKLLARRSNSLLKYITDKSEEIAQINLERKAQKKKKSEDGSKNSRQPAEAGVVESEN
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    APITD1 (NP_954988.1, 1 a.a. ~ 138 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against a full-length recombinant APITD1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: APITD1
Clonality: Monoclonal
Clone: 6F16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human