Anti-N4BP1 Mouse Polyclonal Antibody
Catalog # ABNOH00009683-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NEDD4 binding protein 1
- Antigen symbol:N4BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:9683
- Antigen synonyms:N4BP1|KIAA0615|NEDD4-binding protein 1
- Amino acid number:1 to 896
- Storage buffer:1x PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:N4BP1 (AAI60019.1, 1 a.a. ~ 896 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human N4BP1 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: N4BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human