Anti-TGM1 Rabbit Polyclonal Antibody
ABNOH00007051-D01
: Abnova
- Antibody type:Primary
- Antigen name:Transglutaminase 1
- Antigen symbol:TGM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:7051
- Antigen synonyms:LI|TGASE|ICR2|LI1|KTG|TGK|ARCI1
- Amino acid number:1 to 817
- Storage buffer:No additive
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:TGM1 (NP_000350.1, 1 a.a. ~ 817 a.a) full-length human protein.
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against a full-length human TGM1 protein.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: TGM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse