Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-CK7 Mouse Monoclonal Antibody [clone: CK-7]
  ABNOMAB1442
 :  Abnova
Anti-CK7 Mouse Monoclonal Antibody [clone: CK-7]
  ABNOMAB1442
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Cytokeratin 7
  • Antigen symbol:
    CK7
  • Clonality:
    Monoclonal
  • Clone:
    CK-7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Gene ID:
    3855
  • Antigen synonyms:
    CK7|SCL|K2C7|K7
  • Storage buffer:
    1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified human KRT7.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against native KRT7.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml; Immunohistochemistry (Frozen sections): 2-4 ?g/ml

Type: Primary
Antigen: CK7
Clonality: Monoclonal
Clone: CK-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human