Anti-TRAF2 Rabbit Polyclonal Antibody
ABNOPAB0275
: Abnova
- Antibody type:Primary
- Antigen name:TNF receptor-associated factor 2
- Antigen symbol:TRAF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:7186
- Antigen synonyms:TRAP|TRAP3|TNF receptor-associated factor 2|TRAF2|MGC,45012|E3 ubiquitin-protein ligase TRAF2
- Amino acid number:87 to 106
- Storage buffer:0,05% sodium azide
- Sequence:SSSAFPDNAARREVESLPAV
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 87-106 of human TRAF2.
- Size:50 μl
- Pk:50 µl
Rabbit polyclonal antibody raised against synthetic peptide of TRAF2.
Type: Primary
Antigen: TRAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat