Anti-LDB1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB10257
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:LIM Domain binding 1
- Antigen symbol:LDB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Frog,Chicken,Chimpanzee,Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:16825
- Antigen synonyms:LIM domain-binding protein 1|CLIM 2|LIM Domain Binding 1|NLI|LIM domain binding factor CLIM2|Carboxyl-terminal LIM domain-binding protein 2|ldb1|Carboxyl Terminal LIM Domain Binding 2|CLIM-2|LDB-1|LIM domain-binding factor CLIM2|hLdb1|LDB1_HUMAN|Nuclear LIM Domain Interactor|Nuclear LIM interactorxldb1.
- Amino acid number:361 to 373
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 361-373 of mouse Ldb1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Ldb1.
Recommended Dilutions: ELISA: 1:170000; Western Blot: 1:500-1:3000
Type: Primary
Antigen: LDB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Chimpanzee, Dog, Frog, Human, Mouse, Rat