Anti-RAMP1 Goat Polyclonal Antibody
Catalog # ABNOPAB11575
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Receptor (G Protein-coupled) activity modifying protein 1
- Antigen symbol:RAMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:51801
- Antigen synonyms:9130218E19Rik
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-DYQSKRTEGIV
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to mouse Ramp1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of Ramp1.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: RAMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat